missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-(trifluoromethoxy)benzothiazole, 98%

CAS: 1744-22-5 | C8H5F3N2OS | 234.196 g/mol

Supplier:  Thermo Scientific Chemicals H6132814

 View more versions of this product

Catalog No. AAH6132814


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-6-(trifluoromethoxy)benzothiazole
114°C to 118°C
C8H5F3N2OS
25 g
748759
FTALBRSUTCGOEG-UHFFFAOYSA-N
6-(trifluoromethoxy)-1,3-benzothiazol-2-amine
5070
234.2
Solid
1744-22-5
White to Yellow
MFCD00210213
UN2811
riluzole, rilutek, 2-amino-6-trifluoromethoxy benzothiazole, 6-trifluoromethoxy benzo d thiazol-2-amine, 6-trifluoromethoxy-1,3-benzothiazol-2-amine, riluzolum, riluzol, 2-amino-6-trifluoromethoxybenzothiazole, riluzole hcl, riluzol inn-spanish
C1=CC2=C(C=C1OC(F)(F)F)SC(=N2)N
234.196
CHEBI:8863
98%

Chemical Identifiers

1744-22-5
234.196
FTALBRSUTCGOEG-UHFFFAOYSA-N
5070
6-(trifluoromethoxy)-1,3-benzothiazol-2-amine
C8H5F3N2OS
MFCD00210213
riluzole, rilutek, 2-amino-6-trifluoromethoxy benzothiazole, 6-trifluoromethoxy benzo d thiazol-2-amine, 6-trifluoromethoxy-1,3-benzothiazol-2-amine, riluzolum, riluzol, 2-amino-6-trifluoromethoxybenzothiazole, riluzole hcl, riluzol inn-spanish
CHEBI:8863
C1=CC2=C(C=C1OC(F)(F)F)SC(=N2)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P310-P330-P501c

H300

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : DL2830000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only