missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-(trifluoromethoxy)benzothiazole, 98%

Catalog No. AAH6132806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6132806 5 g
AAH6132814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6132806 Supplier Thermo Scientific Chemicals Supplier No. H6132806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1744-22-5 | C8H5F3N2OS | 234.196 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1744-22-5
Molecular Formula C8H5F3N2OS
Molecular Weight (g/mol) 234.196
MDL Number MFCD00210213
InChI Key FTALBRSUTCGOEG-UHFFFAOYSA-N
Synonym riluzole, rilutek, 2-amino-6-trifluoromethoxy benzothiazole, 6-trifluoromethoxy benzo d thiazol-2-amine, 6-trifluoromethoxy-1,3-benzothiazol-2-amine, riluzolum, riluzol, 2-amino-6-trifluoromethoxybenzothiazole, riluzole hcl, riluzol inn-spanish
PubChem CID 5070
ChEBI CHEBI:8863
IUPAC Name 6-(trifluoromethoxy)-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C=C1OC(F)(F)F)SC(=N2)N

Specifications

Melting Point 114°C to 118°C
Color White to Yellow
Quantity 5 g
UN Number UN2811
Beilstein 748759
Formula Weight 234.2
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Amino-6-(trifluoromethoxy)benzothiazole
Hazard Category H300
Hazard Statement GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber DL2830000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only