missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-(tert-butoxycarbonylamino)pyridine 97.0+%, TCI America™
$175.22 - $594.63
Chemical Identifiers
| CAS | 322690-31-3 |
|---|---|
| Molecular Formula | C10H15N3O2 |
| Molecular Weight (g/mol) | 209.249 |
| MDL Number | MFCD09037865 |
| InChI Key | KEKAANXUQSMDMK-UHFFFAOYSA-N |
| Synonym | 2-Amino-6-(Boc-amino)pyridine |
| PubChem CID | 45092083 |
| IUPAC Name | tert-butyl N-(6-aminopyridin-2-yl)carbamate |
| SMILES | CC(C)(C)OC(=O)NC1=CC=CC(=N1)N |
Chemical Identifiers
| 322690-31-3 | |
| 209.249 | |
| KEKAANXUQSMDMK-UHFFFAOYSA-N | |
| 45092083 | |
| CC(C)(C)OC(=O)NC1=CC=CC(=N1)N |
| C10H15N3O2 | |
| MFCD09037865 | |
| 2-Amino-6-(Boc-amino)pyridine | |
| tert-butyl N-(6-aminopyridin-2-yl)carbamate |
Specifications
| 322690-31-3 | |
| White-Yellow | |
| MFCD09037865 | |
| 2-Amino-6-(Boc-amino)pyridine | |
| CC(C)(C)OC(=O)NC1=CC=CC(=N1)N | |
| 209.249 | |
| 209.25 | |
| Crystalline Powder |
| 127°C | |
| C10H15N3O2 | |
| 1 g | |
| KEKAANXUQSMDMK-UHFFFAOYSA-N | |
| tert-butyl N-(6-aminopyridin-2-yl)carbamate | |
| 45092083 | |
| ≥97.0% (T) | |
| 2-Amino-6-(tert-butoxycarbonylamino)pyridine |
Safety and Handling
TSCA : No