CAS RN 322690-31-3
IUPAC Name:
tert-butyl N-(6-aminopyridin-2-yl)carbamate
Synonyms:
tert-butyl N-(6-aminopyridin-2-yl)carbamate
Molecular Weight (g/mol):
209.25
Molecular Formula:
C10H15N3O2
InChi Key:
KEKAANXUQSMDMK-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NC1=CC=CC(N)=N1
1
–
1
of
1
results
1
2-Amino-6-(tert-butoxycarbonylamino)pyridine 97.0+%, TCI America™
CAS: 322690-31-3 Molecular Formula: C10H15N3O2 Molecular Weight (g/mol): 209.249 MDL Number: MFCD09037865 InChI Key: KEKAANXUQSMDMK-UHFFFAOYSA-N Synonym: 2-Amino-6-(Boc-amino)pyridine PubChem CID: 45092083 IUPAC Name: tert-butyl N-(6-aminopyridin-2-yl)carbamate SMILES: CC(C)(C)OC(=O)NC1=CC=CC(=N1)N