missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-(tert-butoxycarbonylamino)pyridine 97.0+%, TCI America™

Catalog No. A27085G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27085G 5 g
A27081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27085G Supplier TCI America Supplier No. A27085G

Chemical Identifiers

CAS 322690-31-3
Molecular Formula C10H15N3O2
Molecular Weight (g/mol) 209.249
MDL Number MFCD09037865
InChI Key KEKAANXUQSMDMK-UHFFFAOYSA-N
Synonym 2-Amino-6-(Boc-amino)pyridine
PubChem CID 45092083
IUPAC Name tert-butyl N-(6-aminopyridin-2-yl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=CC(=N1)N

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 5 g
Formula Weight 209.25
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-(tert-butoxycarbonylamino)pyridine