missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-methylpyrazine 97.0+%, TCI America™
$61.51 - $173.37
Chemical Identifiers
| CAS | 5521-56-2 |
|---|---|
| Molecular Formula | C5H7N3 |
| Molecular Weight (g/mol) | 109.132 |
| MDL Number | MFCD08235193 |
| InChI Key | UAOOJJPSCLNTOP-UHFFFAOYSA-N |
| Synonym | 2-amino-6-methylpyrazine, pyrazinamine, 6-methyl, 6-methylpyrazine-2-ylamine, 6-methyl-2-pyrazinamine, 2-pyrazinamine,6-methyl, 2-amino-6-methyl pyrazine, 6-methyl-pyrazin-2-ylamine, acmc-1am75, 6-methyl-pyrazinamine, 6-methylpyrazin-2-ylamine |
| PubChem CID | 12236939 |
| IUPAC Name | 6-methylpyrazin-2-amine |
| SMILES | CC1=CN=CC(=N1)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A2992200MG
|
TCI America
A2992200MG |
200 mg |
Each for $61.51
|
|
|||||
|
A29921G
|
TCI America
A29921G |
1 g |
Each for $173.37
|
|
|||||
Chemical Identifiers
| 5521-56-2 | |
| 109.132 | |
| UAOOJJPSCLNTOP-UHFFFAOYSA-N | |
| 12236939 | |
| CC1=CN=CC(=N1)N |
| C5H7N3 | |
| MFCD08235193 | |
| 2-amino-6-methylpyrazine, pyrazinamine, 6-methyl, 6-methylpyrazine-2-ylamine, 6-methyl-2-pyrazinamine, 2-pyrazinamine,6-methyl, 2-amino-6-methyl pyrazine, 6-methyl-pyrazin-2-ylamine, acmc-1am75, 6-methyl-pyrazinamine, 6-methylpyrazin-2-ylamine | |
| 6-methylpyrazin-2-amine |
Specifications
| 5521-56-2 | |
| C5H7N3 | |
| 200 mg | |
| UAOOJJPSCLNTOP-UHFFFAOYSA-N | |
| 6-methylpyrazin-2-amine | |
| 12236939 | |
| ≥97.0% (GC) | |
| 2-Amino-6-methylpyrazine |
| 124°C | |
| MFCD08235193 | |
| 2-amino-6-methylpyrazine, pyrazinamine, 6-methyl, 6-methylpyrazine-2-ylamine, 6-methyl-2-pyrazinamine, 2-pyrazinamine,6-methyl, 2-amino-6-methyl pyrazine, 6-methyl-pyrazin-2-ylamine, acmc-1am75, 6-methyl-pyrazinamine, 6-methylpyrazin-2-ylamine | |
| CC1=CN=CC(=N1)N | |
| 109.132 | |
| 109.13 | |
| Crystalline Powder |
Safety and Handling
TSCA : No