missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methylpyrazine 97.0+%, TCI America™

Catalog No. a2992200mg
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
200 mg
Catalog No. Quantity
A2992200MG 200 mg
A29921G 1 g
2 options

Catalog No. A2992200MG

Supplier: TCI America A2992200MG

Only null left

Chemical Identifiers

CAS 5521-56-2
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.132
MDL Number MFCD08235193
InChI Key UAOOJJPSCLNTOP-UHFFFAOYSA-N
Synonym 2-amino-6-methylpyrazine, pyrazinamine, 6-methyl, 6-methylpyrazine-2-ylamine, 6-methyl-2-pyrazinamine, 2-pyrazinamine,6-methyl, 2-amino-6-methyl pyrazine, 6-methyl-pyrazin-2-ylamine, acmc-1am75, 6-methyl-pyrazinamine, 6-methylpyrazin-2-ylamine
PubChem CID 12236939
IUPAC Name 6-methylpyrazin-2-amine
SMILES CC1=CN=CC(=N1)N

Specifications

Melting Point 124°C
Quantity 200 mg
Formula Weight 109.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-methylpyrazine