missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7117176
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
A09965G 5 g
A099625G 25 g
2 options

Catalog No. A09965G

Supplier: TCI America A09965G

Only null left

Chemical Identifiers

CAS 4389-50-8
Molecular Formula C8H8NO2
Molecular Weight (g/mol) 150.16
MDL Number MFCD00007809
InChI Key XHYVBIXKORFHFM-UHFFFAOYSA-M
Synonym 6-methylanthranilic acid, 6-amino-o-toluic acid, benzoic acid, 2-amino-6-methyl, 2-amino-6-methyl-benzoic acid, 6-methylanthranilate, pubchem4957, acmc-209jvt, 6-methyl anthranilic acid, 3-amino-2-carboxytoluene, 2-carboxy-3-methylaniline
PubChem CID 151210
IUPAC Name 2-amino-6-methylbenzoate
SMILES CC1=CC=CC(N)=C1C([O-])=O

Specifications

Melting Point 124°C
Quantity 5 g
Formula Weight 151.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-methylbenzoic Acid