missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methyl-4(3H)-quinazolone, 96%, Thermo Scientific Chemicals

Catalog No. p-7046489
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1455803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1455803 Supplier Thermo Scientific Chemicals Supplier No. L1455803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50440-82-9 | Molecular Formula: C9H9N3O | Molecular Weight: 175.191 g/mol

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 50440-82-9
Molecular Formula C9H9N3O
Molecular Weight (g/mol) 175.191
MDL Number MFCD00236015
InChI Key DJYDUJAKTAWKTN-UHFFFAOYSA-N
Synonym 2-amino-6-methylquinazolin-4 3h-one, 2-amino-6-methyl-4 3h-quinazolone, 2-amino-6-methyl-3h-quinazolin-4-one, 2-amino-6-methyl-4 3h-quinazolinone, acmc-1aogs, chembl7222, 2-amino-4-hydroxy-6-methylquinazoline, 2-amino-6-methylquinazolin-4 1h-one, 6-methyl-2-aminoquinazolin-4 3h-one, 2-amino-6-methyl-3-hydroquinazolin-4-one
PubChem CID 4151444
IUPAC Name 2-amino-6-methyl-1H-quinazolin-4-one
SMILES CC1=CC2=C(C=C1)NC(=NC2=O)N

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 609421
Solubility Information Slightly soluble in water.
Formula Weight 175.19
Percent Purity 96%
Chemical Name or Material 2-Amino-6-methyl-4(3H)-quinazolone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only