accessibility menu, dialog, popup

UserName

CAS RN 50440-82-9

IUPAC Name: 2-amino-6-methyl-1,4-dihydroquinazolin-4-one
Synonyms: 2-amino-6-methyl-1H-quinazolin-4-one
Molecular Weight (g/mol): 175.19
Molecular Formula: C9H9N3O
InChi Key: DJYDUJAKTAWKTN-UHFFFAOYSA-N
SMILES: CC1=CC=C2NC(N)=NC(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results