missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methyl-3-nitropyridine 98.0+%, TCI America™

Catalog No. p-7117845
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23341G 1 g
A23345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23341G Supplier TCI America Supplier No. A23341G
Only null left

Chemical Identifiers

CAS 21901-29-1
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
MDL Number MFCD00047443
InChI Key LCJXSRQGDONHRK-UHFFFAOYSA-N
Synonym 2-amino-3-nitro-6-picoline, 2-amino-6-methyl-3-nitropyridine, 2-amino-3-nitro-6-methylpyridine, 6-amino-5-nitro-2-picoline, 6-methyl-3-nitro-pyridin-2-ylamine, 6-methyl-3-nitro-2-pyridinamine, 2-pyridinamine, 6-methyl-3-nitro, 6-methyl-3-nitro-2-pyridylamine, 6-methyl-3-nitropyridin-2-amin, 2-amino-3-nitro-6-picoline 2-amino-6-methyl-3-nitropyridine
PubChem CID 226028
IUPAC Name 6-methyl-3-nitropyridin-2-amine
SMILES CC1=NC(=C(C=C1)[N+](=O)[O-])N

Specifications

Melting Point 156°C
Color Yellow
Quantity 1 g
Formula Weight 153.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-methyl-3-nitropyridine