accessibility menu, dialog, popup

UserName

CAS RN 21901-29-1

IUPAC Name: 2-amino-6-methyl-3-nitropyridin-1-ium
Synonyms: 2-amino-6-methyl-3-nitropyridin-1-ium
Molecular Weight (g/mol): 154.15
Molecular Formula: C6H8N3O2
InChi Key: LCJXSRQGDONHRK-UHFFFAOYSA-O
SMILES: CC1=CC=C(C(N)=[NH+]1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)