missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methyl-3-nitropyridine, 98%

Catalog No. aah6172906
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAH6172906 5 g
AAH6172914 25 g
2 options

Catalog No. AAH6172906

Supplier: Thermo Scientific Chemicals H6172906

Only null left

CAS: 21901-29-1 | C6H7N3O2 | 153.141 g/mol

2-Amino-6-methyl-3-nitropyridine used as a intermediate in organic synthesis and used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-6-methyl-3-nitropyridine used as a intermediate in organic synthesis and used in medicine.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry and well-ventilated place. Keep container tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 21901-29-1
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
MDL Number MFCD00047443
InChI Key LCJXSRQGDONHRK-UHFFFAOYSA-N
Synonym 2-amino-3-nitro-6-picoline, 2-amino-6-methyl-3-nitropyridine, 2-amino-3-nitro-6-methylpyridine, 6-amino-5-nitro-2-picoline, 6-methyl-3-nitro-pyridin-2-ylamine, 6-methyl-3-nitro-2-pyridinamine, 2-pyridinamine, 6-methyl-3-nitro, 6-methyl-3-nitro-2-pyridylamine, 6-methyl-3-nitropyridin-2-amin, 2-amino-3-nitro-6-picoline 2-amino-6-methyl-3-nitropyridine
PubChem CID 226028
IUPAC Name 6-methyl-3-nitropyridin-2-amine
SMILES CC1=NC(=C(C=C1)[N+](=O)[O-])N

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 153.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Amino-6-methyl-3-nitropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only