missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-hydroxy-8-mercaptopurine, 97%
Description
Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.
Spécifications
Spécifications
| Nom ou substance chimique | 2-Amino-6-hydroxy-8-mercaptopurine |
| Nom Remarque | 97% |
| CAS % minimum | 96.0 |
| CAS % maximal | 100.0 |
| Point de fusion | >300,0 °C |
| Couleur | Jaune |
| Spectre infrarouge | Authentique |
| Fourchette de pourcentages d’essai | 97% |
| Emballage | Bouteille en verre |
| Quantité | 25 g |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement: Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| TSCA | TSCA |
RUO – Usage réservé à la recherche