accessibility menu, dialog, popup

UserName

CAS RN 28128-40-7

IUPAC Name: 2-amino-8-sulfanylidene-6,7,8,9-tetrahydro-3H-purin-6-one
Synonyms: 2-amino-8-sulfanylidene-7,9-dihydro-3H-purin-6-one
Molecular Weight (g/mol): 183.19
Molecular Formula: C5H5N5OS
InChi Key: JHEKNTQSGTVPAO-UHFFFAOYSA-N
SMILES: NC1=NC(=O)C2=C(NC(=S)N2)N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results