missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-fluoropyridine 98.0+%, TCI America™

Catalog No. A28795G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28795G 5 g
A28791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28795G Supplier TCI America Supplier No. A28795G

Chemical Identifiers

CAS 1597-32-6
Molecular Formula C5H5FN2
Molecular Weight (g/mol) 112.107
MDL Number MFCD04114200
InChI Key UZALKVXCOUSWSL-UHFFFAOYSA-N
Synonym 2-amino-6-fluoropyridine, 6-fluoro-pyridin-2-ylamine, 6-fluoro-2-pyridylamine, 6-fluoropyridin-2-ylamine, 6-fluoro-2-pyridinamine, 2-pyridinamine, 6-fluoro, acmc-209dk6, 6-fluoro-pyridin-2-ylamin, ksc497g8r, 6-amino-2-fluoropyridine
PubChem CID 2761399
IUPAC Name 6-fluoropyridin-2-amine
SMILES C1=CC(=NC(=C1)F)N

Specifications

Melting Point 59°C
Color White-Yellow
Quantity 5 g
Formula Weight 112.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-fluoropyridine