accessibility menu, dialog, popup

UserName

CAS RN 1597-32-6

NFH2N

CAS RN 1597-32-6

IUPAC Name: 6-fluoropyridin-2-amine
Synonyms: 6-fluoropyridin-2-amine
Molecular Weight (g/mol): 112.11
Molecular Formula: C5H5FN2
InChi Key: UZALKVXCOUSWSL-UHFFFAOYSA-N
SMILES: NC1=NC(F)=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)