missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-fluoropyridine, 97%

Catalog No. p-7030590
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
AAH3062903 1 g
AAH3062906 5 g
2 options

Catalog No. AAH3062903

Supplier: Thermo Scientific Chemicals H3062903

Only null left

CAS: 1597-32-6 | C5H5FN2 | 112.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1597-32-6
Molecular Formula C5H5FN2
Molecular Weight (g/mol) 112.107
MDL Number MFCD04114200
InChI Key UZALKVXCOUSWSL-UHFFFAOYSA-N
Synonym 2-amino-6-fluoropyridine, 6-fluoro-pyridin-2-ylamine, 6-fluoro-2-pyridylamine, 6-fluoropyridin-2-ylamine, 6-fluoro-2-pyridinamine, 2-pyridinamine, 6-fluoro, acmc-209dk6, 6-fluoro-pyridin-2-ylamin, ksc497g8r, 6-amino-2-fluoropyridine
PubChem CID 2761399
IUPAC Name 6-fluoropyridin-2-amine
SMILES C1=CC(=NC(=C1)F)N

Specifications

Melting Point 58°C to 62°C
Odor Characteristic
Quantity 1 g
UN Number UN2811
Formula Weight 112.11
Percent Purity 97%
Chemical Name or Material 2-Amino-6-fluoropyridine
Hazard Category H301-H312-H315-H319-H335
Hazard Statement GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only