missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-fluorophenol, 97%

Catalog No. AAH6662603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6662603 1 g
AAH6662606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6662603 Supplier Thermo Scientific Chemicals Supplier No. H6662603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 53981-25-2 | C6H6FNO | 127.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53981-25-2
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.118
MDL Number MFCD08532465
InChI Key MDIAVSZFIQWYST-UHFFFAOYSA-N
Synonym 6-fluoro-2-aminophenol, 3-fluoro-2-hydroxyaniline, 2-amino-6-fluoro-phenol, phenol, 2-amino-6-fluoro, pubchem10659, acmc-209lbm, 2-amino-6-flourophenol, phenol,2-amino-6-fluoro
PubChem CID 643562
IUPAC Name 2-amino-6-fluorophenol
SMILES C1=CC(=C(C(=C1)F)O)N

Specifications

Quantity 1 g
UN Number UN2811
Beilstein 3603077
Formula Weight 127.12
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-6-fluorophenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only