missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-ethylpyridine, 97%

Catalog No. AAA1914609
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1914609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1914609 Supplier Thermo Scientific Chemicals Supplier No. A1914609

CAS: 21717-29-3 | C7H10N2 | 122.171 g/mol

The action of iodine on 2-amino-6-ethylpyridine in an alkaline medium gives 2-amino-5-iodo-6-ethylpyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The action of iodine on 2-amino-6-ethylpyridine in an alkaline medium gives 2-amino-5-iodo-6-ethylpyridine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 21717-29-3
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00218265
InChI Key JXKAUUVMXZIJNZ-UHFFFAOYSA-N
Synonym 2-amino-6-ethylpyridine, 6-ethyl-2-pyridinamine, 2-pyridinamine, 6-ethyl, 6-ethyl-2-pyridylamine, acmc-1clku, 2-pyridinamine,6-ethyl, 2-amino-6-ethyl-pyridine, 6-ethyl-pyridin-2-ylamine
PubChem CID 89021
IUPAC Name 6-ethylpyridin-2-amine
SMILES CCC1=NC(=CC=C1)N

Specifications

Odor Unpleasant
Refractive Index 1.568
Quantity 10 g
UN Number UN2810
Solubility Information Slightly soluble in water.
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 2-Amino-6-ethylpyridine
Hazard Category H302+H332-H311-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 244-540-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only