accessibility menu, dialog, popup

UserName

H3CH2NHN

CAS RN 21717-29-3

IUPAC Name: 2-amino-6-ethylpyridin-1-ium
Synonyms: 2-amino-6-ethylpyridin-1-ium
Molecular Weight (g/mol): 123.18
Molecular Formula: C7H11N2
InChi Key: JXKAUUVMXZIJNZ-UHFFFAOYSA-O
SMILES: CCC1=CC=CC(N)=[NH+]1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results