missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-ethoxybenzothiazole 97.0+%, TCI America™

Catalog No. A071625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A071625G 25 g
A0716500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A071625G Supplier TCI America Supplier No. A071625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 94-45-1
Molecular Formula C9H10N2OS
Molecular Weight (g/mol) 194.252
MDL Number MFCD00005788
InChI Key KOYJWFGMEBETBU-UHFFFAOYSA-N
Synonym 2-amino-6-ethoxybenzothiazole, 2-benzothiazolamine, 6-ethoxy, 6-ethoxy-2-aminobenzothiazole, 6-ethoxy-benzothiazol-2-ylamine, 6-ethoxy-2-benzothiazolamine, 6-ethoxybenzothiazol-2-ylamine, benzothiazole, 2-amino-6-ethoxy, unii-d621hz191d, amino-6-ethoxybenzothiazole, 2, 2-benzothiazolamine, 6-ethoxy-9ci
PubChem CID 7192
IUPAC Name 6-ethoxy-1,3-benzothiazol-2-amine
SMILES CCOC1=CC2=C(C=C1)N=C(S2)N

Specifications

Melting Point 162°C
Quantity 25 g
Formula Weight 194.25
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-ethoxybenzothiazole
EINECSNumber (5)-0239
TSCA No