missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-chloropyridine, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 45644-21-1 |
|---|---|
| Molecular Formula | C5H5ClN2 |
| Molecular Weight (g/mol) | 128.56 |
| MDL Number | MFCD00234068 |
| InChI Key | OBYJTLDIQBWBHM-UHFFFAOYSA-N |
| Synonym | 2-amino-6-chloropyridine, 2-chloro-6-aminopyridine, 2-pyridinamine, 6-chloro, 6-chloro-2-pyridinamine, 6-chloro-pyridin-2-ylamine, pyridine, 2-amino-6-chloro, 6-chloropyridin-2-ylamine, 6-chloro-2-pyridylamine, 6-amino-2-chloropyridine, 2-amino-6-chloropyridine 6-chloropyridin-2-ylamine |
| PubChem CID | 206246 |
| IUPAC Name | 6-chloropyridin-2-amine |
| SMILES | C1=CC(=NC(=C1)Cl)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC429500010
|
N/A | 1g | Glass bottle |
N/A
|
|
||||
|
AC429500050
|
N/A | 5g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 45644-21-1 | |
| 128.56 | |
| OBYJTLDIQBWBHM-UHFFFAOYSA-N | |
| 206246 | |
| C1=CC(=NC(=C1)Cl)N |
| C5H5ClN2 | |
| MFCD00234068 | |
| 2-amino-6-chloropyridine, 2-chloro-6-aminopyridine, 2-pyridinamine, 6-chloro, 6-chloro-2-pyridinamine, 6-chloro-pyridin-2-ylamine, pyridine, 2-amino-6-chloro, 6-chloropyridin-2-ylamine, 6-chloro-2-pyridylamine, 6-amino-2-chloropyridine, 2-amino-6-chloropyridine 6-chloropyridin-2-ylamine | |
| 6-chloropyridin-2-amine |
Specifications
| 45644-21-1 | |
| 100.0 | |
| 96% min. (GC) | |
| MFCD00234068 | |
| OBYJTLDIQBWBHM-UHFFFAOYSA-N | |
| 6-chloropyridin-2-amine | |
| 206246 | |
| 97% |
| 96.0 | |
| Authentic | |
| C5H5ClN2 | |
| 2-amino-6-chloropyridine, 2-chloro-6-aminopyridine, 2-pyridinamine, 6-chloro, 6-chloro-2-pyridinamine, 6-chloro-pyridin-2-ylamine, pyridine, 2-amino-6-chloro, 6-chloropyridin-2-ylamine, 6-chloro-2-pyridylamine, 6-amino-2-chloropyridine, 2-amino-6-chloropyridine 6-chloropyridin-2-ylamine | |
| C1=CC(=NC(=C1)Cl)N | |
| 128.56 | |
| 128.56 | |
| 2-Amino-6-chloropyridine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning
RUO – Research Use Only