Learn More
2-Amino-6-chloropyrazine, 95%
CAS: 33332-28-4 | C4H4ClN3 | 129.547 g/mol
Supplier: Thermo Scientific Chemicals H6193414
Description
2-Amino-6-chloropyrazine is used in the preparation of A2B adenosine receptor antagonists and other biologically active compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-6-chloropyrazine is used in the preparation of A2B adenosine receptor antagonists and other biologically active compounds.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications
| 2-Amino-6-chloropyrazine | |
| C4H4ClN3 | |
| 25 g | |
| Soluble in dimethyl sulfoxide and methanol. | |
| C1=C(N=C(C=N1)Cl)N | |
| 129.547 | |
| 129.55 | |
| Solid |
| 33332-28-4 | |
| MFCD00055024 | |
| 2-amino-6-chloropyrazine, 2-chloro-6-aminopyrazine, 2-pyrazinamine, 6-chloro, pyrazinamine, 6-chloro, 6-chloro-pyrazin-2-ylamine, 6-chloro-2-pyrazinamine, 6-chloropyrazin-2-ylamine, 2-amino-6-chloro-pyrazine, 6-chloropyrazine-2-ylamine, 6-amino-2-chloropyrazine | |
| JTPXVCKCLBROOJ-UHFFFAOYSA-N | |
| 6-chloropyrazin-2-amine | |
| 118458 | |
| 95% |
Chemical Identifiers
| 33332-28-4 | |
| 129.547 | |
| JTPXVCKCLBROOJ-UHFFFAOYSA-N | |
| 118458 | |
| C1=C(N=C(C=N1)Cl)N |
| C4H4ClN3 | |
| MFCD00055024 | |
| 2-amino-6-chloropyrazine, 2-chloro-6-aminopyrazine, 2-pyrazinamine, 6-chloro, pyrazinamine, 6-chloro, 6-chloro-pyrazin-2-ylamine, 6-chloro-2-pyrazinamine, 6-chloropyrazin-2-ylamine, 2-amino-6-chloro-pyrazine, 6-chloropyrazine-2-ylamine, 6-amino-2-chloropyrazine | |
| 6-chloropyrazin-2-amine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only