missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloropyrazine, 95%

Catalog No. p-7054059
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6193414 25 g
AAH6193422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6193414 Supplier Thermo Scientific Chemicals Supplier No. H6193414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33332-28-4 | C4H4ClN3 | 129.547 g/mol

2-Amino-6-chloropyrazine is used in the preparation of A2B adenosine receptor antagonists and other biologically active compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-6-chloropyrazine is used in the preparation of A2B adenosine receptor antagonists and other biologically active compounds.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 33332-28-4
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.547
MDL Number MFCD00055024
InChI Key JTPXVCKCLBROOJ-UHFFFAOYSA-N
Synonym 2-amino-6-chloropyrazine, 2-chloro-6-aminopyrazine, 2-pyrazinamine, 6-chloro, pyrazinamine, 6-chloro, 6-chloro-pyrazin-2-ylamine, 6-chloro-2-pyrazinamine, 6-chloropyrazin-2-ylamine, 2-amino-6-chloro-pyrazine, 6-chloropyrazine-2-ylamine, 6-amino-2-chloropyrazine
PubChem CID 118458
IUPAC Name 6-chloropyrazin-2-amine
SMILES C1=C(N=C(C=N1)Cl)N

Specifications

Quantity 25 g
Solubility Information Soluble in dimethyl sulfoxide and methanol.
Formula Weight 129.55
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-6-chloropyrazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only