missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chlorobenzonitrile, 98%

CAS: 6575-11-7 | C7H5ClN2 | 152.58 g/mol

$49.07 - $155.88

Chemical Identifiers

CAS 6575-11-7
MDL Number MFCD00015546
PubChem CID 81056
IUPAC Name 2-amino-6-chlorobenzonitrile
SMILES C1=CC(=C(C(=C1)Cl)C#N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0688303
View Documents
Thermo Scientific Chemicals
L0688303
1 g
Each for $49.07
Only null left
 
AAL0688306
View Documents
Thermo Scientific Chemicals
L0688306
5 g
Each for $155.88
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6575-11-7
81056
C1=CC(=C(C(=C1)Cl)C#N)N
MFCD00015546
2-amino-6-chlorobenzonitrile

Specifications

133°C to 137°C
1 g
MFCD00015546
2717090
QEIFSLUFHRCVQL-UHFFFAOYSA-N
152.58
2-Amino-6-chlorobenzonitrile
6575-11-7
C15H15BrClN2O4P
UN3439
6-Chloroanthranilonitrile
433.62
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 229-501-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only