missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chlorobenzonitrile, 98%

CAS: 6575-11-7 | C7H5ClN2 | 152.58 g/mol

Supplier:  Thermo Scientific Chemicals L0688306

 View more versions of this product

Catalog No. AAL0688306


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-6-chlorobenzonitrile
6575-11-7
C7H5ClN2
UN3439
6-chloroanthranilonitrile, benzonitrile, 2-amino-6-chloro, 2-amino-6-chloro-benzonitrile, 3-chloro-2-cyanoaniline, pubchem4614, acmc-1b6zr, 6-amino-2-chlorobenzonitrile, ksc497g6t, 2-amino-6-chloro benzonitrile, benzonitrile,2-amino-6-chloro
C1=CC(=C(C(=C1)Cl)C#N)N
152.58
152.58
133°C to 137°C
5 g
MFCD00015546
2717090
MEJVTQKBWPYBFG-UHFFFAOYSA-N
2-amino-6-chlorobenzonitrile
81056
98%

Chemical Identifiers

6575-11-7
81056
C1=CC(=C(C(=C1)Cl)C#N)N
MFCD00015546
2-amino-6-chlorobenzonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 229-501-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only