Learn More
2-Amino-6-chlorobenzoic acid, 97+%
CAS: 2148-56-3 | C7H6ClNO2 | 171.58 g/mol
Supplier: Thermo Scientific Chemicals A1089009
Description
2-Amino-6-chlorobenzoic acid is used as intermediate for medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-6-chlorobenzoic acid is used as intermediate for medicine.
Solubility
Very soluble in water. Soluble in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and bases.
Specifications
| 2-Amino-6-chlorobenzoic acid | |
| 149°C to 152°C | |
| MFCD00051530 | |
| 2804041 | |
| Very soluble in water. Soluble in methanol. | |
| C1=CC(=C(C(=C1)Cl)C(=O)O)N | |
| 171.58 | |
| 171.58 |
| 2148-56-3 | |
| C7H6ClNO2 | |
| 10 g | |
| 6-chloroanthranilic acid, benzoic acid, 2-amino-6-chloro, 2-amino-6-chloro-benzoic acid, 6-chloranthranilsaure, pubchem9876, acmc-1cngq, 6-chloro anthranilic acid, 2-carboxy-3-chloroaniline, 2amino-6-chlorobenzoic acid, ksc201s6b | |
| SZCPTRGBOVXVCA-UHFFFAOYSA-N | |
| 2-amino-6-chlorobenzoic acid | |
| 75071 | |
| 98% |
Chemical Identifiers
| 2148-56-3 | |
| 171.58 | |
| SZCPTRGBOVXVCA-UHFFFAOYSA-N | |
| 75071 | |
| C1=CC(=C(C(=C1)Cl)C(=O)O)N |
| C7H6ClNO2 | |
| MFCD00051530 | |
| 6-chloroanthranilic acid, benzoic acid, 2-amino-6-chloro, 2-amino-6-chloro-benzoic acid, 6-chloranthranilsaure, pubchem9876, acmc-1cngq, 6-chloro anthranilic acid, 2-carboxy-3-chloroaniline, 2amino-6-chlorobenzoic acid, ksc201s6b | |
| 2-amino-6-chlorobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 218-416-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only