missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chlorobenzoic acid, 97+%

Catalog No. AAA1089006
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1089006 5 g
AAA1089009 10 g
AAA1089014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1089006 Supplier Thermo Scientific Chemicals Supplier No. A1089006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2148-56-3 | C7H6ClNO2 | 171.58 g/mol

2-Amino-6-chlorobenzoic acid is used as intermediate for medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-6-chlorobenzoic acid is used as intermediate for medicine.

Solubility
Very soluble in water. Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 2148-56-3
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00051530
InChI Key SZCPTRGBOVXVCA-UHFFFAOYSA-N
Synonym 6-chloroanthranilic acid, benzoic acid, 2-amino-6-chloro, 2-amino-6-chloro-benzoic acid, 6-chloranthranilsaure, pubchem9876, acmc-1cngq, 6-chloro anthranilic acid, 2-carboxy-3-chloroaniline, 2amino-6-chlorobenzoic acid, ksc201s6b
PubChem CID 75071
IUPAC Name 2-amino-6-chlorobenzoic acid
SMILES C1=CC(=C(C(=C1)Cl)C(=O)O)N

Specifications

Melting Point 149°C to 152°C
Quantity 5 g
Beilstein 2804041
Solubility Information Very soluble in water. Soluble in methanol.
Formula Weight 171.58
Percent Purity 98%
Chemical Name or Material 2-Amino-6-chlorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-416-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only