missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-chloro-4-nitrophenol 98.0+%, TCI America™
$169.63 - $581.59
Chemical Identifiers
| CAS | 6358-09-4 |
|---|---|
| Molecular Formula | C6H6Cl2N2O3 |
| Molecular Weight (g/mol) | 225.03 |
| MDL Number | MFCD00035767 |
| InChI Key | HXKQPFKBBNRPEK-UHFFFAOYSA-N |
| Synonym | 3-Chloro-2-hydroxy-5-nitroaniline |
| PubChem CID | 4679699 |
| SMILES | C1=C(C=C(C(=C1Cl)O)N)[N+](=O)[O-] |
Chemical Identifiers
| 6358-09-4 | |
| 225.03 | |
| HXKQPFKBBNRPEK-UHFFFAOYSA-N | |
| 4679699 |
| C6H6Cl2N2O3 | |
| MFCD00035767 | |
| 3-Chloro-2-hydroxy-5-nitroaniline | |
| C1=C(C=C(C(=C1Cl)O)N)[N+](=O)[O-] |
Specifications
| 6358-09-4 | |
| C6H6Cl2N2O3 | |
| 5 g | |
| HXKQPFKBBNRPEK-UHFFFAOYSA-N | |
| 225.03 | |
| 188.57 | |
| Crystalline Powder |
| 162°C | |
| MFCD00035767 | |
| 3-Chloro-2-hydroxy-5-nitroaniline | |
| C1=C(C=C(C(=C1Cl)O)N)[N+](=O)[O-] | |
| 4679699 | |
| ≥98.0% (T) | |
| 2-Amino-6-chloro-4-nitrophenol |
Safety and Handling
EINECSNumber : (3)-0939
RTECSNumber : ST5743000
TSCA : Yes