missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloro-4-nitrophenol 98.0+%, TCI America™

Catalog No. A22455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22455G 5 g
A224525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22455G Supplier TCI America Supplier No. A22455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6358-09-4
Molecular Formula C6H6Cl2N2O3
Molecular Weight (g/mol) 225.03
MDL Number MFCD00035767
InChI Key HXKQPFKBBNRPEK-UHFFFAOYSA-N
Synonym 3-Chloro-2-hydroxy-5-nitroaniline
PubChem CID 4679699
SMILES C1=C(C=C(C(=C1Cl)O)N)[N+](=O)[O-]

Specifications

Melting Point 162°C
Quantity 5 g
Formula Weight 188.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-chloro-4-nitrophenol
EINECSNumber (3)-0939
RTECSNumber ST5743000
TSCA Yes