missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-chloro-3-formylchromone 98.0+%, TCI America™
$95.06 - $285.21
Chemical Identifiers
| CAS | 68301-77-9 |
|---|---|
| Molecular Formula | C10H6ClNO3 |
| Molecular Weight (g/mol) | 223.61 |
| MDL Number | MFCD00139140 |
| InChI Key | GCFVPLUXLQEQJX-UHFFFAOYSA-N |
| Synonym | 2-Amino-6-chlorochromone-3-carboxaldehyde, 2-Amino-6-chloro-4-oxo-4H-1-benzopyran-3-carboxaldehyde |
| PubChem CID | 689030 |
| IUPAC Name | 2-amino-6-chloro-4-oxo-4H-chromene-3-carbaldehyde |
| SMILES | NC1=C(C=O)C(=O)C2=CC(Cl)=CC=C2O1 |
Chemical Identifiers
| 68301-77-9 | |
| 223.61 | |
| GCFVPLUXLQEQJX-UHFFFAOYSA-N | |
| 689030 | |
| NC1=C(C=O)C(=O)C2=CC(Cl)=CC=C2O1 |
| C10H6ClNO3 | |
| MFCD00139140 | |
| 2-Amino-6-chlorochromone-3-carboxaldehyde, 2-Amino-6-chloro-4-oxo-4H-1-benzopyran-3-carboxaldehyde | |
| 2-amino-6-chloro-4-oxo-4H-chromene-3-carbaldehyde |
Specifications
| 68301-77-9 | |
| C10H6ClNO3 | |
| 1 g | |
| GCFVPLUXLQEQJX-UHFFFAOYSA-N | |
| 2-amino-6-chloro-4-oxo-4H-chromene-3-carbaldehyde | |
| 689030 | |
| ≥98.0% (HPLC,N) | |
| 2-Amino-6-chloro-3-formylchromone |
| 290°C | |
| MFCD00139140 | |
| 2-Amino-6-chlorochromone-3-carboxaldehyde, 2-Amino-6-chloro-4-oxo-4H-1-benzopyran-3-carboxaldehyde | |
| NC1=C(C=O)C(=O)C2=CC(Cl)=CC=C2O1 | |
| 223.61 | |
| 223.61 | |
| Crystalline Powder |
Safety and Handling
TSCA : No