accessibility menu, dialog, popup

UserName

CAS RN 68301-77-9

H2NOOClO

CAS RN 68301-77-9

IUPAC Name: 2-amino-6-chloro-4-oxo-4H-chromene-3-carbaldehyde
Synonyms: 2-amino-6-chloro-4-oxochromene-3-carbaldehyde
Molecular Weight (g/mol): 223.61
Molecular Formula: C10H6ClNO3
InChi Key: GCFVPLUXLQEQJX-UHFFFAOYSA-N
SMILES: NC1=C(C=O)C(=O)C2=CC(Cl)=CC=C2O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results