missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-(trifluoromethyl)pyridine 97.0+%, TCI America™

Catalog No. p-7117664
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19685G 5 g
A196825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19685G Supplier TCI America Supplier No. A19685G

Chemical Identifiers

CAS 74784-70-6
Molecular Formula C6H5F3N2
Molecular Weight (g/mol) 162.115
MDL Number MFCD00042164
InChI Key RSGVKIIEIXOMPY-UHFFFAOYSA-N
Synonym 5-trifluoromethyl pyridin-2-amine, 2-amino-5-trifluoromethyl pyridine, 2-amino-5-trifluoromethylpyridine, 5-trifluoromethyl-2-pyridinamine, 5-trifluoromethyl-pyridin-2-ylamine, 2-pyridinamine, 5-trifluoromethyl, 5-trifluoromethyl-2-pyridylamine, 2-amine-5-trifluoromethylpyridine, 5-trifluormethyl pyridin-2-amin
PubChem CID 735862
IUPAC Name 5-(trifluoromethyl)pyridin-2-amine
SMILES C1=CC(=NC=C1C(F)(F)F)N

Specifications

Melting Point 47°C
Color White-Yellow
Quantity 5 g
Formula Weight 162.12
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-(trifluoromethyl)pyridine