Learn More
2-Amino-5-(trifluoromethyl)benzoic acid, 95%
CAS: 83265-53-6 | C8H6F3NO2 | 205.136 g/mol
Supplier: Thermo Scientific Chemicals H6414103
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Amino-5-(trifluoromethyl)benzoic acid | |
| 141°C to 146°C (decomposition) | |
| C8H6F3NO2 | |
| 1 g | |
| GLCQUPLYYXSPQB-UHFFFAOYSA-N | |
| 2-amino-5-(trifluoromethyl)benzoic acid | |
| 10465535 | |
| 95% |
| 83265-53-6 | |
| >110°C (230°F) | |
| MFCD01569537 | |
| 2-amino-5-trifluoromethyl benzoic acid, 2-amino-5-trifluoromethyl-benzoic acid, 5-trifluoromethylanthranilic acid, 5-trifluoromethyl anthranilic acid, benzoic acid, 2-amino-5-trifluoromethyl, 2-amino-5-trifluoromethyl-benzoicacid, pubchem12904, 4-amino-3-carboxybenzotrifluoride, 2-carboxy-4-trifluoromethyl aniline, 2-amino-5-trifluoromethyl benzoicacid | |
| C1=CC(=C(C=C1C(F)(F)F)C(=O)O)N | |
| 205.136 | |
| 205.14 | |
| Solid |
Chemical Identifiers
| 83265-53-6 | |
| 205.136 | |
| GLCQUPLYYXSPQB-UHFFFAOYSA-N | |
| 10465535 | |
| C1=CC(=C(C=C1C(F)(F)F)C(=O)O)N |
| C8H6F3NO2 | |
| MFCD01569537 | |
| 2-amino-5-trifluoromethyl benzoic acid, 2-amino-5-trifluoromethyl-benzoic acid, 5-trifluoromethylanthranilic acid, 5-trifluoromethyl anthranilic acid, benzoic acid, 2-amino-5-trifluoromethyl, 2-amino-5-trifluoromethyl-benzoicacid, pubchem12904, 4-amino-3-carboxybenzotrifluoride, 2-carboxy-4-trifluoromethyl aniline, 2-amino-5-trifluoromethyl benzoicacid | |
| 2-amino-5-(trifluoromethyl)benzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only