missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-(trifluoromethoxy)benzoic acid, 98%

Catalog No. AAH3291606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3291606 5 g
AAH3291603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3291606 Supplier Thermo Scientific Chemicals Supplier No. H3291606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 83265-56-9 | C8H6F3NO3 | 221.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83265-56-9
Molecular Formula C8H6F3NO3
Molecular Weight (g/mol) 221.135
MDL Number MFCD00176651
InChI Key UXNGDCBPIGOZFO-UHFFFAOYSA-N
Synonym 2-amino-5-trifluoromethoxy benzoic acid, 2-amino-5-trifluoromethoxy-benzoic acid, 5-trifluoromethoxyanthranilic acid, 2-amino-5-trifluoromethoxy benzoicacid, benzoic acid, 2-amino-5-trifluoromethoxy, pubchem12913, acmc-20a4df, ksc496m3d, uxngdcbpigozfo-uhfffaoysa, 5-trifluoromethoxy anthranilic acid
PubChem CID 11085349
IUPAC Name 2-amino-5-(trifluoromethoxy)benzoic acid
SMILES C1=CC(=C(C=C1OC(F)(F)F)C(=O)O)N

Specifications

Quantity 5 g
Formula Weight 221.14
Percent Purity 98%
Chemical Name or Material 2-Amino-5-(trifluoromethoxy)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only