missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-tert-butyl-1,3,4-thiadiazole, 97%

Catalog No. AAL0611606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0611606 5 g
AAL0611603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0611606 Supplier Thermo Scientific Chemicals Supplier No. L0611606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39222-73-6 | C6H11N3S | 157.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39222-73-6
Molecular Formula C6H11N3S
Molecular Weight (g/mol) 157.24
MDL Number MFCD00040403
InChI Key ICXDPEFCLDSXLI-UHFFFAOYSA-N
Synonym 2-amino-5-tert-butyl-1,3,4-thiadiazole, buthidazolamine, 5-tert-butyl-1,3,4-thiadiazol-2-amine, 1,3,4-thiadiazol-2-amine, 5-1,1-dimethylethyl, unii-x53j1t94sn, 5-tert-butyl-1,3,4 thiadiazol-2-ylamine, 5-tert-butyl-1,3,4-thiadiazole-2-ylamine, acmc-20aidm, enamine_005678, 5-t-butyl-2-amino1,3,4-thiadiazole
PubChem CID 170170
IUPAC Name 5-tert-butyl-1,3,4-thiadiazol-2-amine
SMILES CC(C)(C)C1=NN=C(N)S1

Specifications

Melting Point 181°C to 185°C
Quantity 5 g
Beilstein 119853
Formula Weight 157.24
Percent Purity 97%
Physical Form Crystals or Powder or Crystalline Powder
Chemical Name or Material 2-Amino-5-tert-butyl-1,3,4-thiadiazole
EINECSNumber 254-364-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only