accessibility menu, dialog, popup

UserName

CH3H3CCH3SNH2NN

CAS RN 39222-73-6

IUPAC Name: 5-tert-butyl-1,3,4-thiadiazol-2-amine
Synonyms: 5-tert-butyl-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 157.24
Molecular Formula: C6H11N3S
InChi Key: ICXDPEFCLDSXLI-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=NN=C(N)S1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)