missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-phenylpyrazine 98.0+%, TCI America™
$147.25 - $443.65
Chemical Identifiers
| CAS | 13535-13-2 |
|---|---|
| Molecular Formula | C10H9N3 |
| Molecular Weight (g/mol) | 171.20 |
| MDL Number | MFCD01646266 |
| InChI Key | KJAKXVBZQBPPOB-UHFFFAOYSA-N |
| Synonym | 2-amino-5-phenylpyrazine, pyrazinamine, 5-phenyl, pyrazine, 2-amino-5-phenyl, acmc-1bpgx, 5-phenyl-2-pyrazinamine, 5-phenyl-2-aminopyrazine, 2-pyrazinamine,5-phenyl, 5-phenylpyrazine-2-ylamine, 5-phenyl-2-pyrazinamine # |
| PubChem CID | 598734 |
| IUPAC Name | 5-phenylpyrazin-2-amine |
| SMILES | NC1=NC=C(N=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 13535-13-2 | |
| 171.20 | |
| KJAKXVBZQBPPOB-UHFFFAOYSA-N | |
| 598734 | |
| NC1=NC=C(N=C1)C1=CC=CC=C1 |
| C10H9N3 | |
| MFCD01646266 | |
| 2-amino-5-phenylpyrazine, pyrazinamine, 5-phenyl, pyrazine, 2-amino-5-phenyl, acmc-1bpgx, 5-phenyl-2-pyrazinamine, 5-phenyl-2-aminopyrazine, 2-pyrazinamine,5-phenyl, 5-phenylpyrazine-2-ylamine, 5-phenyl-2-pyrazinamine # | |
| 5-phenylpyrazin-2-amine |
Specifications
| 13535-13-2 | |
| C10H9N3 | |
| 1 g | |
| KJAKXVBZQBPPOB-UHFFFAOYSA-N | |
| 5-phenylpyrazin-2-amine | |
| 598734 | |
| ≥98.0% (GC,T) | |
| 2-Amino-5-phenylpyrazine |
| 146°C | |
| MFCD01646266 | |
| 2-amino-5-phenylpyrazine, pyrazinamine, 5-phenyl, pyrazine, 2-amino-5-phenyl, acmc-1bpgx, 5-phenyl-2-pyrazinamine, 5-phenyl-2-aminopyrazine, 2-pyrazinamine,5-phenyl, 5-phenylpyrazine-2-ylamine, 5-phenyl-2-pyrazinamine # | |
| NC1=NC=C(N=C1)C1=CC=CC=C1 | |
| 171.20 | |
| 171.20 | |
| Crystalline Powder |
Safety and Handling
TSCA : No