CAS RN 13535-13-2
IUPAC Name:
5-phenylpyrazin-2-amine
Synonyms:
5-phenylpyrazin-2-amine
Molecular Weight (g/mol):
171.2
Molecular Formula:
C10H9N3
InChi Key:
KJAKXVBZQBPPOB-UHFFFAOYSA-N
SMILES:
NC1=NC=C(N=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-Amino-5-phenylpyrazine 98.0+%, TCI America™
CAS: 13535-13-2 Molecular Formula: C10H9N3 Molecular Weight (g/mol): 171.20 MDL Number: MFCD01646266 InChI Key: KJAKXVBZQBPPOB-UHFFFAOYSA-N Synonym: 2-amino-5-phenylpyrazine,pyrazinamine, 5-phenyl,pyrazine, 2-amino-5-phenyl,acmc-1bpgx,5-phenyl-2-pyrazinamine,5-phenyl-2-aminopyrazine,2-pyrazinamine,5-phenyl,5-phenylpyrazine-2-ylamine,5-phenyl-2-pyrazinamine # PubChem CID: 598734 IUPAC Name: 5-phenylpyrazin-2-amine SMILES: NC1=NC=C(N=C1)C1=CC=CC=C1