missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-phenyl-1,3,4-thiadiazol 98.0+%, TCI America™

Catalog No. A30021G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A30021G Supplier TCI America Supplier No. A30021G

Chemical Identifiers

CAS 2002-03-1
Molecular Formula C8H7N3S
Molecular Weight (g/mol) 177.23
MDL Number MFCD00205278
InChI Key UHZHEOAEJRHUBW-UHFFFAOYSA-N
Synonym 5-Phenyl-1,3,4-thiadiazol-2-amine
PubChem CID 219408
IUPAC Name 5-phenyl-1,3,4-thiadiazol-2-amine
SMILES C1=CC=C(C=C1)C2=NN=C(S2)N

Specifications

Melting Point 227°C
Quantity 1 g
Formula Weight 177.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-phenyl-1,3,4-thiadiazol