accessibility menu, dialog, popup

UserName

NH2SNN

CAS RN 2002-03-1

IUPAC Name: 5-phenyl-1,3,4-thiadiazol-2-amine
Synonyms: 5-phenyl-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 177.23
Molecular Formula: C8H7N3S
InChi Key: UHZHEOAEJRHUBW-UHFFFAOYSA-N
SMILES: NC1=NN=C(S1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)