missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-phenyl-1,3,4-oxadiazole 97.0+%, TCI America™
Supplier: TCI America A30745G
Specifications
| 2-Amino-5-phenyl-1,3,4-oxadiazole | |
| 211°C | |
| C8H7N3O | |
| 5 g | |
| NC1=NN=C(O1)C1=CC=CC=C1 | |
| 161.16 | |
| 161.16 | |
| Crystalline Powder |
| 1612-76-6 | |
| Yellow | |
| MFCD00126383 | |
| CQSFYCBGVMWPCM-UHFFFAOYSA-N | |
| 5-phenyl-1,3,4-oxadiazol-2-amine | |
| 15363 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 1612-76-6 | |
| 161.16 | |
| CQSFYCBGVMWPCM-UHFFFAOYSA-N | |
| 5-phenyl-1,3,4-oxadiazol-2-amine |
| C8H7N3O | |
| MFCD00126383 | |
| 15363 | |
| NC1=NN=C(O1)C1=CC=CC=C1 |
Safety and Handling
TSCA : No