CAS RN 1612-76-6
IUPAC Name:
5-phenyl-1,3,4-oxadiazol-2-amine
Synonyms:
5-phenyl-1,3,4-oxadiazol-2-amine
Molecular Weight (g/mol):
161.16
Molecular Formula:
C8H7N3O
InChi Key:
CQSFYCBGVMWPCM-UHFFFAOYSA-N
SMILES:
NC1=NN=C(O1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-Amino-5-phenyl-1,3,4-oxadiazole 97.0+%, TCI America™
CAS: 1612-76-6 Molecular Formula: C8H7N3O Molecular Weight (g/mol): 161.16 MDL Number: MFCD00126383 InChI Key: CQSFYCBGVMWPCM-UHFFFAOYSA-N PubChem CID: 15363 IUPAC Name: 5-phenyl-1,3,4-oxadiazol-2-amine SMILES: NC1=NN=C(O1)C1=CC=CC=C1