missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-phenyl-1,3,4-oxadiazole 97.0+%, TCI America™

Catalog No. A30741G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30741G 1 g
A30745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30741G Supplier TCI America Supplier No. A30741G

Chemical Identifiers

CAS 1612-76-6
Molecular Formula C8H7N3O
Molecular Weight (g/mol) 161.16
MDL Number MFCD00126383
InChI Key CQSFYCBGVMWPCM-UHFFFAOYSA-N
PubChem CID 15363
IUPAC Name 5-phenyl-1,3,4-oxadiazol-2-amine
SMILES NC1=NN=C(O1)C1=CC=CC=C1

Specifications

Melting Point 211°C
Color Yellow
Quantity 1 g
Formula Weight 161.16
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-phenyl-1,3,4-oxadiazole