missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrophenol 98.0+%, TCI America™

Catalog No. p-7133626
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
N011325G 25 g
N0113500G 500 g
2 options

Catalog No. N011325G

Supplier: TCI America N011325G

Only null left

Chemical Identifiers

CAS 121-88-0
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00007692
InChI Key DOPJTDJKZNWLRB-UHFFFAOYSA-N
Synonym 2-hydroxy-4-nitroaniline, 5-nitro-2-aminophenol, phenol, 2-amino-5-nitro, ursol yellow brown a, rodol yba, 3-nitro-6-aminophenol, 3-hydroxy-4-aminonitrobenzene, 2-amino-5-nitro-phenol, ccris 754, 4-amino-3-hydroxynitrobenzene
PubChem CID 4984721
ChEBI CHEBI:82384
IUPAC Name 2-amino-5-nitrophenol
SMILES C1=CC(=C(C=C1[N+](=O)[O-])O)N

Specifications

Melting Point 204°C
Quantity 25 g
Formula Weight 154.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-nitrophenol
EINECSNumber (3)-0788
RTECSNumber SJ6302500
TSCA Yes