missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrophenol, 95%

Catalog No. AC400902500
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
1 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC400902500 250 g Glass bottle
AC400900250 25 g Glass bottle
AC400900500 50 g Glass bottle
AC400900010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC400902500 Supplier Thermo Scientific Chemicals Supplier No. 400902500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 121-88-0 | C6H6N2O3 | 154.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121-88-0
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00007692
InChI Key DOPJTDJKZNWLRB-UHFFFAOYSA-N
Synonym 2-hydroxy-4-nitroaniline, 5-nitro-2-aminophenol, phenol, 2-amino-5-nitro, ursol yellow brown a, rodol yba, 3-nitro-6-aminophenol, 3-hydroxy-4-aminonitrobenzene, 2-amino-5-nitro-phenol, ccris 754, 4-amino-3-hydroxynitrobenzene
PubChem CID 4984721
ChEBI CHEBI:82384
IUPAC Name 2-amino-5-nitrophenol
SMILES C1=CC(=C(C=C1[N+](=O)[O-])O)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 198.0°C to 202.0°C
Color Brown to Brown-Rust
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 250 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, benzene and diethylether
Formula Weight 154.13
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-nitrophenol, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice
EINECSNumber 204-503-8
RTECSNumber SJ6302500
TSCA TSCA

RUO – Research Use Only