missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrophenol, 95%

Catalog No. AAB2087022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2087022 100 g
AAB2087014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2087022 Supplier Thermo Scientific Chemicals Supplier No. B2087022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 121-88-0 | C6H6N2O3 | 154.125 g/mol

2-Amino-5-nitrophenol has been used as starting material in the synthesis of series of (Z)-2-(substituted aryl)-N-(3-oxo-4-(substituted carbamothioyl)-3,4-dihydro-2H-benzo[b][1,4]oxazin-7-yl) hydrazine carboxamides, as semi permanent (nonoxidative) hair colorant and as toner in permanent (oxidative) hair dye products, in synthesis of series of 7-benzylamino-2H-1,4-benzoxazin-3(4H)-ones, anticonvulsant agents. Also used as a reactant for, diazotation and coupling reactions, preparation of biologically and pharmacologically active molecules. It is also applied in hair dyes and to make azo dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-nitrophenol has been used as starting material in the synthesis of series of (Z)-2-(substituted aryl)-N-(3-oxo-4-(substituted carbamothioyl)-3,4-dihydro-2H-benzo[b][1,4]oxazin-7-yl) hydrazine carboxamides, as semi permanent (nonoxidative) hair colorant and as toner in permanent (oxidative) hair dye products, in synthesis of series of 7-benzylamino-2H-1,4-benzoxazin-3(4H)-ones, anticonvulsant agents. Also used as a reactant for, diazotation and coupling reactions, preparation of biologically and pharmacologically active molecules. It is also applied in hair dyes and to make azo dyes.

Solubility
Insoluble in water. Solubility in methanol gives very faint turbidity. Soluble in alcohol, benzene, acetone, and diethyl ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 121-88-0
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00007692
InChI Key DOPJTDJKZNWLRB-UHFFFAOYSA-N
Synonym 2-hydroxy-4-nitroaniline, 5-nitro-2-aminophenol, phenol, 2-amino-5-nitro, ursol yellow brown a, rodol yba, 3-nitro-6-aminophenol, 3-hydroxy-4-aminonitrobenzene, 2-amino-5-nitro-phenol, ccris 754, 4-amino-3-hydroxynitrobenzene
PubChem CID 4984721
ChEBI CHEBI:82384
IUPAC Name 2-amino-5-nitrophenol
SMILES C1=CC(=C(C=C1[N+](=O)[O-])O)N

Specifications

Melting Point ∼200°C (decomposition)
Quantity 100 g
Beilstein 972974
Solubility Information Insoluble in water. Solubility in methanol gives very faint turbidity. Soluble in alcohol,benzene,acetone,and diethyl ether.
Formula Weight 154.13
Percent Purity 96%
Chemical Name or Material 2-Amino-5-nitrophenol
Hazard Category H302-H317-H319
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P363-P501c
EINECSNumber 204-503-8
RTECSNumber SJ6302500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only