missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrobenzotrifluoride 98.0+%, TCI America™

Catalog No. A08051G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08051G 1 g
A08055G 5 g
A080525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A08051G Supplier TCI America Supplier No. A08051G

Chemical Identifiers

CAS 121-01-7
Molecular Formula C7H5F3N2O2
Molecular Weight (g/mol) 206.124
MDL Number MFCD00007365
InChI Key HOTZLWVITTVZGY-UHFFFAOYSA-N
Synonym 2-amino-5-nitrobenzotrifluoride, 4-nitro-2-trifluoromethyl aniline, 2-amino-5-nitro-trifluorobenzene, 4-nitro-alpha,alpha,alpha-trifluoro-o-toluidine, 4-nitro-2-trifluoromethyl benzenamine, 4-nitro-a,a,a-trifluoro-o-toluidine, benzenamine, 4-nitro-2-trifluoromethyl, 5-nitro-2-aminobenzotrifluoride, 2-trifluoromethyl-4-nitroaniline, a,a,a-trifluoro-4-nitro-o-toluidine
PubChem CID 67128
IUPAC Name 4-nitro-2-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)N

Specifications

Melting Point 91°C
Quantity 1 g
Formula Weight 206.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-nitrobenzotrifluoride