missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-nitrobenzonitrile 98.0+%, TCI America™
Supplier: TCI America A131425G
Specifications
| 2-Amino-5-nitrobenzonitrile | |
| 210°C | |
| MFCD00007362 | |
| 5-nitroanthranilonitrile, 2-cyano-4-nitroaniline, benzonitrile, 2-amino-5-nitro, 2-kyan-4-nitroanilin, unii-sez79wra1j, 2-amino-5-nitro-benzonitrile, ccris 9092, 2-kyan-4-nitroanilin czech, aniline, 2-cyano-4-nitro, sez79wra1j | |
| C1=CC(=C(C=C1[N+](=O)[O-])C#N)N | |
| 163.136 | |
| 163.14 | |
| Crystalline Powder |
| 17420-30-3 | |
| C7H5N3O2 | |
| 25 g | |
| MGCGMYPNXAFGFA-UHFFFAOYSA-N | |
| 2-amino-5-nitrobenzonitrile | |
| 28532 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 17420-30-3 | |
| 163.136 | |
| MGCGMYPNXAFGFA-UHFFFAOYSA-N | |
| 28532 | |
| C1=CC(=C(C=C1[N+](=O)[O-])C#N)N |
| C7H5N3O2 | |
| MFCD00007362 | |
| 5-nitroanthranilonitrile, 2-cyano-4-nitroaniline, benzonitrile, 2-amino-5-nitro, 2-kyan-4-nitroanilin, unii-sez79wra1j, 2-amino-5-nitro-benzonitrile, ccris 9092, 2-kyan-4-nitroanilin czech, aniline, 2-cyano-4-nitro, sez79wra1j | |
| 2-amino-5-nitrobenzonitrile |
Safety and Handling
EINECSNumber : (3)-1806
RTECSNumber : BX2500000
TSCA : Yes